Abnova
Manufacturer Code:MAB19372
Catalog # 89-338-876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | D23 |
Immunogen | Recombinant protein corresponding to amino acids 542-602 at C-terminus of rat Kcna5 |
Conjugate | ATTO 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C |
Mouse Monoclonal Antibody.
Protein Aliases: Kv1 Kv1.5; Potassium (K+) channel protein alpha 5; potassium voltage gated channel, shaker related subfamily, member 5; Potassium voltage-gated channel subfamily A member 5; potassium voltage-gated channel, shaker-related subfamily, member 5; RCK7; RK4; Voltage-gated potassium channel subunit Kv1.5
Gene Aliases: Kcna5; Kv1; Kv1.5
UniProt ID: (Rat) P19024
Entrez Gene ID: (Rat) 25470
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.