Proteintech
Manufacturer Code:142321AP150UL
Catalog # 50-172-9300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Kallikrein 8 Fusion Protein Ag5472 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain Serine protease 1; hK8; hK8 HNP Kallikrein 8 kallikrein related peptidase 8 KLK8 Neuropsin NP NRPN Ovasin PRSS19 Serine protease 19 Serine protease TADG 14 TADG14; kallikrein 8; kallikrein 8 (neuropsin/ovasin); Kallikrein-8; kallikrein-related peptidase 8; mK8; Neuropsin; NP; Ovasin; protease, serine, 19 (neuropsin); Serine protease 19; Serine protease TADG-14; Tumor-associated differentially expressed gene 14 protein
Gene Aliases: BSP1; HNP; KLK8; NP; NRPN; PRSS19; TADG14; UNQ283/PRO322
UniProt ID: (Human) O60259, (Mouse) Q61955
Entrez Gene ID: (Human) 11202, (Rat) 308565, (Mouse) 259277
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.