Invitrogen
Manufacturer Code:PA586830
Catalog # PIPA586830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in Human KLK7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hK7; hSCCE; kallikrein 7 (chymotryptic, stratum corneum); kallikrein related-peptidase 7 (chymotryptic, stratum corneum); Kallikrein-7; Kallikrein-7 hK7 Serine protease 6 Stratum corneum chymotryptic enzyme hSCCE KLK7 PRSS6 SCCE; kallikrein-related peptidase 7; protease, serine, 6; Serine protease 6; signal protein; Stratum corneum chymotryptic enzyme; Thymopsin
Gene Aliases: hK7; KLK7; PRSS6; SCCE
UniProt ID: (Human) P49862, (Mouse) Q91VE3
Entrez Gene ID: (Human) 5650, (Mouse) 23993, (Rat) 292852
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.