Invitrogen
Manufacturer Code:PA588324
Catalog # PIPA588324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human KLK6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kallikrein 6; Kallikrein-6; Neurosin; Protease M; Serine protease 18; Serine protease 9; SP59; Zyme
Gene Aliases: Bssp; hK6; KLK6; Klk7; PRSS18; PRSS9; SP59
UniProt ID: (Human) Q92876
Entrez Gene ID: (Human) 5653
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.