Invitrogen
Manufacturer Code:PA586829
Catalog # PIPA586829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in Human KLK6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain serine protease; kallikrein 29; kallikrein 6; kallikrein 6 (neurosin); kallikrein 6 (neurosin, zyme); Kallikrein-6; Kallikrein-6 Neurosin Protease M SP59 Serine protease 18 Serine protease 9 Zyme KLK6 PRSS18 PRSS9; myelencephalon-specific protease; myofibril-bound protease; Neurosin; Protease M; protease serine 9 (neurosin); protease, serine, 18; protease, serine, 9; Serine protease 18; Serine protease 9; SP59; Zyme
Gene Aliases: AI849898; BSP; Bssp; hK6; Klk29; KLK6; Klk7; MSP; neurosin; PRSS18; PRSS9; SP59
UniProt ID: (Human) Q92876
Entrez Gene ID: (Human) 5653, (Mouse) 19144, (Rat) 29245
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.