Invitrogen
Manufacturer Code:PA581431
Catalog # PIPA581431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse KLK6/Kallikrein 6/Neurosin protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glandular kallikrein K1; KAL-B; kallikrein 6; kallikrein renal/pancreas/salivary; Kallikrein-1; mGK-6; Renal kallikrein; Tissue kallikrein-6; true tissue kallikrein
Gene Aliases: 0610007D04Rik; Kal; Klk-6; Klk1; Klk1b6; Klk6; mGk-6
UniProt ID: (Mouse) P15947
Entrez Gene ID: (Mouse) 16612
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.