Invitrogen
Manufacturer Code:PA519117
Catalog # PIPA519117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KEKPGVYTDVCTHIR) corresponding to the C-terminus amino acids of Klk6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: brain serine protease; kallikrein 29; kallikrein 6; kallikrein-6; Klk6 kallikrein related-peptidase 6 AI849898 Bssp Klk29 Klk7 Prss18 Prss9 neurosin kallikrein 29 kallikrein 6 kallikrein 7 protease M protease serine 18 protease serine 9; myelencephalon-specific protease; myofibril-bound protease; protease M; protease, serine, 18; protease, serine, 9
Gene Aliases: AI849898; BSP; Bssp; Klk29; MSP; neurosin; Prss18; Prss9
Entrez Gene ID: (Mouse) 19144
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.