Proteintech
Manufacturer Code:105142AP150UL
Catalog # 50-172-6369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Kallikrein 5 Fusion Protein Ag0799 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kallikrein 5; Kallikrein 5 Kallikrein like protein 2 kallikrein related peptidase 5 KLK L2 KLK5 KLKL2 SCTE Stratum corneum tryptic enzyme; Kallikrein-5; Kallikrein-like protein 2; kallikrein-related peptidase 5; KLK-L2; Stratum corneum tryptic enzyme
Gene Aliases: KLK-L2; KLK5; KLKL2; SCTE; UNQ570/PRO1132
UniProt ID: (Human) Q9Y337
Entrez Gene ID: (Human) 25818
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.