Invitrogen
Manufacturer Code:PA579567
Catalog # PIPA579567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Kallikrein 2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glandular kallikrein 2; Glandular kallikrein-1; hGK-1; kallikrein 2, prostatic; Kallikrein-2; Kallikrein-2 Glandular kallikrein-1 hGK-1 Tissue kallikrein-2 KLK2; kallikrein-related peptidase 2; Tissue kallikrein-2
Gene Aliases: hGK-1; hK2; KLK2; KLK2A2
UniProt ID: (Human) P20151
Entrez Gene ID: (Human) 3817
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.