Novus Biologicals
Manufacturer Code:NB200139
Catalog # NB200139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Residues 262-277 [IRKYETQQQKWLKGPQ] of the human KLK-L4 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586J1923 EC 3.4.21 EC 3.4.21.- kallikrein 13 Kallikrein-like protein 4 kallikrein-related peptidase 13 KLK-L4kallikrein-13 KLKL4kallikrein-like gene 4; kallikrein 13; Kallikrein-13; kallikrein-like gene 4; Kallikrein-like protein 4; kallikrein-related peptidase 13; KLK-L4
Gene Aliases: KLK-L4; KLK13; KLKL4
UniProt ID: (Human) Q86VI8
Entrez Gene ID: (Human) 26085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.