Novus Biologicals
Manufacturer Code:NBP303531100UL
Catalog # NB095515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 18-150 of human Kallikrein 12 (NP_062544.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686H1078 EC 3.4.21 EC 3.4.21.- kallikrein 12 Kallikrein-like protein 5 kallikrein-related peptidase 12 KLKL5kallikrein-12 KLK-L5MGC42603; kallikrein 12; Kallikrein-12; Kallikrein-like protein 5; kallikrein-related peptidase 12; KLK-L5
Gene Aliases: KLK-L5; KLK12; KLKL5; UNQ669/PRO1303
UniProt ID: (Human) Q9UKR0
Entrez Gene ID: (Human) 43849
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.