R&D Systems
Manufacturer Code:AF3095
Catalog # AF3095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12 Ala18-Asn248 Accession # Q9UKR0 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686H1078 EC 3.4.21 EC 3.4.21.- kallikrein 12 Kallikrein-like protein 5 kallikrein-related peptidase 12 KLK12 KLK-L5 KLKL5kallikrein-12 KLK-L5MGC42603; kallikrein 12; Kallikrein-12; Kallikrein-like protein 5; kallikrein-related peptidase 12; KLK-L5
Gene Aliases: KLK-L5; KLK12; KLKL5; UNQ669/PRO1303
UniProt ID: (Human) Q9UKR0
Entrez Gene ID: (Human) 43849
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.