Novus Biologicals
Manufacturer Code:NBP299288R
Catalog # NB083291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Kallikrein 1 (NP_002248.1 Met 1-Ser 262). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.21 EC 3.4.21.35 glandular kallikrein 1 hK1 kallikrein 1 kallikrein 1 renal/pancreas/salivary kallikrein serine protease 1 kallikrein-1 Kidney/pancreas/salivary gland kallikrein Klk6 KLKR Tissue kallikrein; glandular kallikrein 1; kallikrein 1, renal/pancreas/salivary; kallikrein serine protease 1; Kallikrein-1; Kidney/pancreas/salivary gland kallikrein; Tissue kallikrein
Gene Aliases: hK1; KLK1; Klk6; KLKR
UniProt ID: (Human) P06870
Entrez Gene ID: (Human) 3816
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.