Invitrogen
Manufacturer Code:PA536953
Catalog # PIPA536953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 895-940 of human KALRN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cyclin B1; huntingtin-associated protein interacting protein (duo); Huntingtin-associated protein-interacting protein; Kalirin; Kalirin Huntingtin-associated protein-interacting protein Protein Duo Serine/threonine-protein kinase with Dbl- and pleckstrin homology domain DUET DUO HAPIP TRAD; kalrn; P-CIP10; PAM COOH-terminal interactor protein 10; Protein Duo; serine/threonine kinase with Dbl- and pleckstrin homology domain; serine/threonine kinase with Dbl- and pleckstrin homology domains; Serine/threonine-protein kinase with Dbl- and pleckstrin homology domain
Gene Aliases: 2210407G14Rik; ARHGEF24; AV235988; CHD5; CHDS5; DUET; DUO; E530005C20Rik; Gm539; HAPIP; Kalirin; KALRN; TRAD
UniProt ID: (Human) O60229, (Rat) P97924, (Mouse) A2CG49
Entrez Gene ID: (Human) 8997, (Rat) 84009, (Mouse) 545156
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.