Invitrogen
Manufacturer Code:MA526643
Catalog # PIMA526643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-216 of KYNU produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HAMAP-Rule:MF&lowbar03017}; HAMAP-Rule:MF&lowbar03017} 3713 {ECO:0000255; HAMAP-Rule:MF&lowbar03017} KYNU {ECO:0000255; HAMAP-Rule:MF&lowbar03017} L-kynurenine hydrolase {ECO:0000255; KYNU; Kynureninase; Kynureninase {ECO:0000255; L-kynurenine hydrolase
Gene Aliases: KYNU; KYNUU
UniProt ID: (Human) Q16719
Entrez Gene ID: (Human) 8942
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.