Invitrogen
Manufacturer Code:PA587233
Catalog # PIPA587233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human KYNU. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KYNU; Kynureninase; Kynureninase L-kynurenine hydrolase KYNU; kynureninase {ECO:0000255|HAMAP-Rule:MF_03017}; L-kynurenine hydrolase; L-kynurenine hydrolase {ECO:0000255|HAMAP-Rule:MF_03017}
Gene Aliases: 4432411A05Rik; KYNU; KYNUU
UniProt ID: (Human) Q16719, (Mouse) Q9CXF0, (Rat) P70712
Entrez Gene ID: (Human) 8942, (Mouse) 70789, (Rat) 116682
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.