Invitrogen
Manufacturer Code:PA568935
Catalog # PIPA568935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human KCNAB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hKvbeta2; K(+) channel subunit beta-2; KCNAB2 KCNA2B KCNK2; KCNAB2.1; KCNAB2.2; Kv-beta-2; Kvbeta2.1; Kvbeta2.2; Neuroimmune protein F5; potassium channel; potassium channel Kv beta 2.1 subunit; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2; Voltage-gated potassium channel subunit beta-2
Gene Aliases: AKR6A5; Ckbeta2; F5; HKvbeta2; HKvbeta2.1; HKvbeta2.2; I2rf5; KCNA2B; KCNAB2; Kcnb3; KCNK2; KV-BETA-2; KVB2; Kvbeta2.1
UniProt ID: (Human) Q13303, (Bovine) Q27955, (Mouse) P62482, (Rat) P62483
Entrez Gene ID: (Dog) 489626, (Human) 8514, (Horse) 100052362, (Rabbit) 100328600, (Bovine) 541597, (Mouse) 16498, (Rat) 29738, (Guinea pig) 100726297
Molecular Function:
ion channel
oxidoreductase
potassium channel
reductase
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.