Invitrogen
Manufacturer Code:PA577641
Catalog # PIPA577641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)TPQHHISLKESTAK corresponding to amino acid residues 54-67 of rat KV beta1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K(+) channel subunit beta-1; K+ channel Beta1a chain; Kv-beta-1; KVbeta1 KCNAB1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; Voltage-gated potassium channel subunit beta-1
Gene Aliases: AKR6A3; Akr8a8; hKvb3; hKvBeta3; KCNA1B; KCNAB1; KV-BETA-1; Kvb1; Kvb1.3; Kvbeta1.1; KvBeta3; mKv(beta)1
UniProt ID: (Human) Q14722, (Mouse) P63143, (Rat) P63144
Entrez Gene ID: (Human) 7881, (Mouse) 16497, (Rat) 29737
Molecular Function: ion channel oxidoreductase potassium channel reductase transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.