Invitrogen
Manufacturer Code:PA541019
Catalog # PIPA541019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human KCNAB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K(+) channel subunit beta-1; K+ channel Beta1a chain; Kv-beta-1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel subunit Kv beta 1; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; Potassium voltage-gated channel shaker-related subfamily beta member 1 hKvb3 AKR6A3 KCNA1B Kvb1.3 hKvBeta3 KV-BETA-1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; Voltage-gated potassium channel subunit beta-1
Gene Aliases: AKR6A3; Akr8a8; hKvb3; hKvBeta3; KCNA1B; KCNAB1; KV-BETA-1; Kvb1; Kvb1.3; KVBETA1; Kvbeta1.1; KvBeta3; mKv(beta)1
UniProt ID: (Rabbit) Q9XT31, (Bovine) Q4PJK1, (Human) Q14722, (Mouse) P63143, (Rat) P63144
Entrez Gene ID: (Rabbit) 100328569, (Bovine) 526133, (Dog) 612006, (Horse) 100050434, (Human) 7881, (Guinea pig) 100725410, (Mouse) 16497, (Rat) 29737
Molecular Function: ion channel oxidoreductase potassium channel reductase transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.