Invitrogen
Manufacturer Code:PA541026
Catalog # PIPA541026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KCND3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: potassium channel Kv4.3; potassium channel Kv4.3L; potassium channel Kv4.3M; potassium channel, voltage gated Shal related subfamily D, member 3; potassium ionic channel Kv4.3; potassium voltage gated channel, Shal-related family, member 3; potassium voltage-gated channel long; Potassium voltage-gated channel Shal-related subfamily member 3 KV4.3 KCND3L KCND3S KSHIVB; Potassium voltage-gated channel subfamily D member 3; potassium voltage-gated channel, Shal-related subfamily, member 3; sha1-related potassium channel Kv4.3; voltage-gated K+ channel; Voltage-gated potassium channel subunit Kv4.3
Gene Aliases: AW045978; BRGDA9; KCND3; KCND3L; KCND3S; Kncd3; KSHIVB; KV4.3; SCA19; SCA22
UniProt ID: (Human) Q9UK17, (Rabbit) Q9TTT5, (Mouse) Q9Z0V1, (Rat) Q62897
Entrez Gene ID: (Human) 3752, (Bovine) 539739, (Horse) 100058973, (Rabbit) 100009472, (Dog) 403758, (Mouse) 56543, (Rat) 65195
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.