Invitrogen
Manufacturer Code:PA530942
Catalog # PIPA530942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 462 and 655 of Human Kv4.3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KCND3L KCND3S KSHIVB KV4.3 potassium ionic channel Kv4.3 potassium voltage-gated channel subfamily D member 3 sha1-related potassium channel Kv4.3 voltage-gated K+ channel voltage-gated potassium channel subunit Kv4.3; potassium channel, voltage gated Shal related subfamily D, member 3; potassium ionic channel Kv4.3; potassium voltage-gated channel long; Potassium voltage-gated channel subfamily D member 3; potassium voltage-gated channel, Shal-related subfamily, member 3; sha1-related potassium channel Kv4.3; voltage-gated K+ channel; Voltage-gated potassium channel subunit Kv4.3
Gene Aliases: BRGDA9; KCND3; KCND3L; KCND3S; KSHIVB; KV4.3; SCA19; SCA22
UniProt ID: (Human) Q9UK17
Entrez Gene ID: (Human) 3752
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.