Invitrogen
Manufacturer Code:PA5109257
Catalog # PIPA5109257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within human Kv4.3 440-480 aa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW045978 BOS_3189 BRGDA9 Kcnd3 KCND3L KCND3S Kncd3 KSHIVB Kv4.3 Potassium voltage-gated channel subfamily D member 3 SCA19 SCA22 Voltage-gated potassium channel subunit Kv4.3; potassium channel, voltage gated Shal related subfamily D, member 3; potassium ionic channel Kv4.3; potassium voltage-gated channel long; Potassium voltage-gated channel subfamily D member 3; potassium voltage-gated channel, Shal-related subfamily, member 3; sha1-related potassium channel Kv4.3; voltage-gated K+ channel; Voltage-gated potassium channel subunit Kv4.3
Gene Aliases: BRGDA9; KCND3; KCND3L; KCND3S; KSHIVB; KV4.3; SCA19; SCA22
UniProt ID: (Human) Q9UK17
Entrez Gene ID: (Human) 3752
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.