Invitrogen
Manufacturer Code:PA531435
Catalog # PIPA531435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 221 of Human Kv4.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KV4.2 RK5 potassium voltage-gated channel subfamily D member 2 voltage-gated potassium channel Kv4.2 voltage-gated potassium channel subunit Kv4.2 voltage-sensitive potassium channel; potassium channel, voltage gated Shal related subfamily D, member 2; Potassium voltage-gated channel subfamily D member 2; Voltage-gated potassium channel subunit Kv4.2; voltage-sensitive potassium channel
Gene Aliases: KCND2; KIAA1044; KV4.2; RK5
UniProt ID: (Human) Q9NZV8
Entrez Gene ID: (Human) 3751
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.