Invitrogen
Manufacturer Code:PA130660
Catalog # PIPA130660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide ASGPMPAPPRQERKRTQDALI conjugated to KLH corresponding to amino acids 23-43 of Rat Kv4.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KCND2; Kv4.2 RK5 Shal1; potassium channel alpha subunit Kv4.2; potassium channel Kv4.2; potassium channel, voltage gated Shal related subfamily D, member 2; potassium voltage gated channel, Shal-related family, member 2; Potassium voltage-gated channel subfamily D member 2; potassium voltage-gated channel, Shal-related subfamily, member 2; RK5; Shal1; Voltage-gated potassium channel subunit Kv4.2; voltage-sensitive potassium channel
Gene Aliases: AI839615; AW555701; KCND2; KIAA1044; KV4.2; mKIAA1044; MNCb-7013; R75121; RK5; Shal1
UniProt ID: (Human) Q9NZV8, (Rabbit) P59995, (Rat) Q63881, (Mouse) Q9Z0V2
Entrez Gene ID: (Human) 3751, (Rabbit) 100008841, (Rat) 65180, (Mouse) 16508
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.