Invitrogen
Manufacturer Code:PA564866
Catalog # PIPA564866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human KCND1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: potassium channel, voltage gated Shal related subfamily D, member 1; Potassium voltage-gated channel subfamily D member 1; Potassium voltage-gated channel subfamily D member 1 Voltage-gated potassium channel subunit Kv4.1 KCND1; potassium voltage-gated channel, Shal-related subfamily, member 1; Shal-type potassium channel; Voltage-gated potassium channel subunit Kv4.1
Gene Aliases: KCND1; KV4.1
UniProt ID: (Human) Q9NSA2
Entrez Gene ID: (Human) 3750
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.