Invitrogen
Manufacturer Code:PA537871
Catalog # PIPA537871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-HISNKTVTQASP from the internal region of KCNC3 (aa317-328) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: KSHIIID; potassium voltage gated channel, Shaw-related subfamily, member 3; potassium voltage-gated channel Shaw-related subfamily member 3 KSHIIID Kcr2-3 potassium voltage-gated channel subfamily C member 3 voltage-gated potassium channel subunit Kv3.3; Potassium voltage-gated channel subfamily C member 3; Voltage-gated potassium channel subunit Kv3.3
Gene Aliases: Kcnc3; Kcr2-3; KShIIID; Kv3.3
UniProt ID: (Mouse) Q63959
Entrez Gene ID: (Mouse) 16504, (Rat) 117101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.