Invitrogen
Manufacturer Code:PA577579
Catalog # PIPA577579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide DLGGKRLGIEDAAGLGGPDGK(C) corresponding to amino acid residues 184-204 of rat KV 3.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kcnc2; Potassium channel voltage-gated Shaw-related subfamily C member 2; potassium channel, voltage gated Shaw related subfamily C, member 2; potassium voltage gated channel, Shaw-related subfamily, member 2; Potassium voltage-gated channel subfamily C member 2; potassium voltage-gated channel, Shaw-related subfamily, member 2; Shaw-like potassium channel; Voltage-gated potassium channel Kv3.2; Voltage-gated potassium channel subunit Kv3.2
Gene Aliases: AW047325; B230117I07; KCNC2; KShIIIA; KV3.2
UniProt ID: (Human) Q96PR1, (Rat) P22462, (Mouse) Q14B80
Entrez Gene ID: (Human) 3747, (Rat) 246153, (Mouse) 268345
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.