Invitrogen
Manufacturer Code:PA5111830
Catalog # PIPA5111830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 841-857 of rat KV2.1 (Intracellular C-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_13247 Delayed rectifier potassium channel 1 DRK1 DRK1PC KCB1 KCNB1 Kcr1-1 Kv2.1 mShab Potassium voltage-gated channel subfamily B member 1 Shab Voltage-gated potassium channel subunit Kv2.1; Delayed rectifier potassium channel 1; DRK1; mShab; potassium channel Kv2.1; potassium voltage gated channel, Shab-related subfamily, member 1; Potassium voltage-gated channel subfamily B member 1; Voltage-gated potassium channel subunit Kv2.1
Gene Aliases: DRK1PC; Kcnb1; Kcr1-1; Kv2.1; Shab
UniProt ID: (Mouse) Q03717, (Rat) P15387
Entrez Gene ID: (Mouse) 16500, (Rat) 25736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.