Invitrogen
Manufacturer Code:PA577652
Catalog # PIPA577652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KDPETLLPTNDIHR corresponding to amino acid residues 187- 200 of human KV 1.8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cyclic GMP gated potassium channel; KV1.8; potassium channel, voltage gated shaker related subfamily A, member 10; potassium voltage gated channel, shaker related subfamily, member 8; Potassium voltage-gated channel subfamily A member 10; potassium voltage-gated channel, shaker-related subfamily, member 10; potassium voltage-gated channel, shaker-related subfamily, member 8; Voltage-gated potassium channel subunit Kv1.8
Gene Aliases: Gm1962; Kcn1; KCNA10; Kcna8; Kv1.8
UniProt ID: (Human) Q16322, (Mouse) B2RQA1
Entrez Gene ID: (Human) 3744, (Rat) 295360, (Mouse) 242151
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.