Invitrogen
Manufacturer Code:PA577573
Catalog # PIPA577573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GST fusion protein with sequence HRETDHEEQAALKEEQGIQRRESGLDTGGQRKVSCSKASFHKTGGPLEST DSIRRGSCPLEKCHLKAKSNVDLRRSLYALCLDTS RETDL corresponding to amino acid residues 513-602 of mouse KV 1.5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac potassium channel; HPCN1; insulinoma and islet potassium channel; Kcna5; KV1-5; Potassium (K+) channel protein alpha 5; potassium channel 1; potassium channel, voltage gated shaker related subfamily A, member 5; potassium voltage gated channel, shaker related subfamily, member 5; Potassium voltage-gated channel subfamily A member 5; potassium voltage-gated channel, shaker-related subfamily, member 5; RCK7; RK4; Voltage-gated potassium channel HK2; voltage-gated potassium channel protein Kv1.5; Voltage-gated potassium channel subunit Kv1.5
Gene Aliases: ATFB7; HCK1; HK2; HPCN1; KCNA5; Kv1; KV1.5; PCN1
UniProt ID: (Human) P22460, (Rat) P19024, (Mouse) Q61762
Entrez Gene ID: (Human) 3741, (Rat) 25470, (Mouse) 16493
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.