Invitrogen
Manufacturer Code:PA570493
Catalog # PIPA570493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human KRTAP1-5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High sulfur keratin-associated protein 1.5; high sulfur protein B2F; keratin associated protein 1-5; keratin associated protein 1.5; Keratin-associated protein 1-5; Keratin-associated protein 1.5; KRTAP1-5 KAP1.5 KRTAP1.5
Gene Aliases: 2310043L02Rik; AI592983; KAP1.5; KRTAP1-5; KRTAP1.5
UniProt ID: (Human) Q9BYS1
Entrez Gene ID: (Human) 83895, (Rat) 497995, (Mouse) 69664
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.