Invitrogen
Manufacturer Code:PA5114142
Catalog # PIPA5114142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KRT75 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4732468K03Rik AA589387 BOS_5011 CK-75 cytokeratin KRT2-6HF cytokeratin type II Cytokeratin-75 hK6hf K6HF K75 Kb18 keratin 75 keratin 75 type II keratin 75-like Keratin type II cytoskeletal 75 Keratin-6 hair follicle Keratin-75 keratinocyte associated protein 1 Krt2-6hf KRT75 Krtcap1 LOC100347139 mK6hf PFB type II keratin Kb18 type II keratin-18 Type II keratin-K6hf Type-II keratin Kb18; CK-75; cytokeratin type II; Cytokeratin-75; hK6hf; K75; keratin 75, type II; Keratin, type II cytoskeletal 75; Keratin-6 hair follicle; Keratin-75; type II keratin-18; Type II keratin-K6hf; Type-II keratin Kb18
Gene Aliases: K6HF; KB18; KRT75; PFB
UniProt ID: (Human) B4DQU4
Entrez Gene ID: (Human) 9119
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.