Proteintech
Manufacturer Code:204491AP150UL
Catalog # 50-173-2897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KRT34 Fusion Protein Ag14295 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ha 4 HA4 Hair keratin type I Ha4 hHa4 HKA4 K34 keratin 34 Keratin type I cuticular Ha4 KRT34 KRTHA4; Hair keratin, type I Ha4; hard keratin, type I, 4; K34; keratin 34, type I; keratin complex 1, acidic, gene 4; keratin, hair, acidic, 4; Keratin, type I cuticular Ha4; Keratin-34; krt34
Gene Aliases: 4733401E01Rik; Ha-4; Ha2; HA4; HHA4; HKA4; Krt1-4; KRT34; KRTHA4
UniProt ID: (Human) O76011, (Mouse) Q9D646
Entrez Gene ID: (Human) 3885, (Mouse) 16672
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.