OriGene
Manufacturer Code:CF500015
Catalog # 50-165-9291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 69-372 of human CK18 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 46 protein; CK-18; CK-18 CYK18 K18; cytokeratin 18; Cytokeratin-18; K18; keratin 18, type I; Keratin, type I cytoskeletal 18; Keratin-18
Gene Aliases: CK-18; CYK18; K18; KRT18; PIG46
UniProt ID: (Human) P05783
Entrez Gene ID: (Human) 3875
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.