Proteintech
Manufacturer Code:120631AP150UL
Catalog # 50-559-444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KRAS Fusion Protein Ag2700 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C K RAS c Ki ras GTPase KRas K Ras 2 K RAS2A K RAS2B K RAS4A K RAS4B KI RAS KRAS KRAS1 KRAS2 KRAS2B speicifc Krasa Krasb NS3 RASK2; c-K-ras; c-K-ras2 protein; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; cellular c-Ki-ras2 proto-oncogene; GTPase KRas; GTPase KRas, N-terminally processed; K-Ras 2; K-ras p21 protein; Ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); oncogene KRAS2; p21 protein; PR310 c-K-ras oncogene; transforming protein p21; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog
Gene Aliases: AI929937; C-K-RAS; c-Ki-ras; CFC2; K-ras; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS; Kras-2; KRAS1; KRAS2; NS; NS3; p21; p21B; RALD; ras; RASK2
UniProt ID: (Human) P01116, (Rat) P08644, (Mouse) P32883
Entrez Gene ID: (Human) 3845, (Rat) 24525, (Mouse) 16653
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.