Proteintech
Manufacturer Code:161561AP150UL
Catalog # 50-173-0760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C K RAS c Ki ras GTPase KRas K Ras 2 K RAS2A K RAS2B K RAS4A K RAS4B KI RAS KRAS KRAS1 KRAS2 KRAS2A KRAS2B speicifc KRAS4A Krasa Krasb NS3 RASK2; c-K-ras; c-Ki-ras; c-Kirsten-ras protein; cellular c-Ki-ras2 proto-oncogene; GTPase KRas; GTPase KRas, N-terminally processed; K-Ras 2; K-ras p21 protein; Ki-Ras; Kirsten rat sarcoma viral oncogene homolog; oncogene KRAS2; PR310 c-K-ras oncogene; transforming protein p21; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog
Gene Aliases: C-K-RAS; CFC2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS; KRAS1; KRAS2; NS; NS3; RALD; RASK2
UniProt ID: (Human) P01116
Entrez Gene ID: (Human) 3845
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.