Invitrogen
Manufacturer Code:PA518450
Catalog # PIPA518450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RRSKTNKAKTLAT) corresponding to the C-terminus amino acids of KPNB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: importin 1; importin 90; Importin beta; importin beta-1 subunit; Importin subunit beta-1; Importin-90; karyopherin (importin) beta 1; Karyopherin subunit beta-1; karyopherin,beta 1; KPNB1 karyopherin (importin) beta 1 HGNC:6400 IMB1 IPOB Impnb MGC2155 MGC2156 MGC2157 NTF97 importin 90 importin beta-1 subunit karyopherin beta 1 nuclear factor p97 KapB1 IPO1; Nuclear factor p97; Pore targeting complex 97 kDa subunit; PTAC97; SCG
Gene Aliases: AA409963; IMB1; Impnb; IPO1; IPOB; KPNB1; NTF97
UniProt ID: (Human) Q14974, (Rat) P52296, (Mouse) Q6GTI5
Entrez Gene ID: (Human) 3837, (Rat) 24917, (Mouse) 16211
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.