Abnova
Manufacturer Code:PAB6909
Catalog # 89-132-790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human KPNB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of KPNB1.
For Research Use Only
Protein Aliases: IMB1; Impnb; importin 1; importin 90; importin beta-1 subunit; Importin subunit beta-1; Importin-90; IPO1; IPOB; karyopherin (importin) beta 1; Karyopherin subunit beta-1; MGC2155; MGC2156; MGC2157; NTF97; Nuclear factor p97; Pore targeting complex 97 kDa subunit; PTAC97
Gene Aliases: IMB1; Impnb; IPO1; IPOB; KPNB1; NTF97
UniProt ID: (Human) Q14974
Entrez Gene ID: (Human) 3837
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.