Invitrogen
Manufacturer Code:PA570789
Catalog # PIPA570789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human KPNA7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 8; Importin subunit alpha-8; karyopherin 7; karyopherin alpha 7 (importin alpha 8); Karyopherin subunit alpha-7; KPNA7 Karyopherin alpha nucleoprotein interactor 7 Importin alpha 8 IPOA8
Gene Aliases: IPOA8; KPNA7
UniProt ID: (Pig) C6K7I2, (Human) A9QM74, (Bovine) C1JZ66
Entrez Gene ID: (Pig) 100302505, (Human) 402569, (Bovine) 789280, (Dog) 489863
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.