Invitrogen
Manufacturer Code:PA596654
Catalog # PIPA596654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 277-536 of human KPNA6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 7 subunit; Importin alpha-S2; Importin subunit alpha-7; importin-alpha-S2; karyopherin (importin) alpha 5; karyopherin alpha 6 (importin alpha 7); Karyopherin subunit alpha-6
Gene Aliases: IPOA7; Kpna5; KPNA6; KPNA7; NPI-2
UniProt ID: (Human) O60684, (Mouse) O35345
Entrez Gene ID: (Human) 23633, (Mouse) 16650
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.