Novus Biologicals
Manufacturer Code:NBP30491920UL
Catalog # NB093832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 277-536 of human KPNA6 (NP_036448.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 7 subunit; importin alpha 7 subunit importin subunit alpha-7 importin-alpha-S2 IPOA7FLJ11249 karyopherin alpha 6 (importin alpha 7) Karyopherin subunit alpha-6 KPNA7 MGC17918; Importin subunit alpha-7; importin-alpha-S2; karyopherin alpha 6 (importin alpha 7); Karyopherin subunit alpha-6
Gene Aliases: IPOA7; KPNA6; KPNA7
UniProt ID: (Human) O60684
Entrez Gene ID: (Human) 23633
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.