Abnova
Manufacturer Code:PAB19318
Catalog # 89-120-851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 13 amino acids near N-terminus of human KPNA6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of KPNA6.
For Research Use Only
Protein Aliases: FLJ11249; importin alpha 7 subunit; Importin subunit alpha-7; importin-alpha-S2; IPOA7; karyopherin alpha 6 (importin alpha 7); Karyopherin subunit alpha-6; KPNA7; MGC17918
Gene Aliases: IPOA7; KPNA6; KPNA7
UniProt ID: (Human) O60684
Entrez Gene ID: (Human) 23633
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.