Invitrogen
Manufacturer Code:PA544199
Catalog # PIPA544199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human KPNA5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 6; Importin subunit alpha-6; Importin subunit alpha-6-like protein; karyopherin alpha 5 (importin alpha 6); Karyopherin alpha 5 (importin alpha 6) IPOA6 SRP6; Karyopherin subunit alpha-5
Gene Aliases: im:7151662; IPOA6; KPNA5; SRP6; zgc:110662
UniProt ID: (Human) O15131, (Rat) Q56R16, (Zebrafish) Q503E9
Entrez Gene ID: (Pig) 397220, (Dog) 484094, (Human) 3841, (Rabbit) 100343572, (Horse) 100067045, (Rat) 294392, (Guinea pig) 100718275, (Zebrafish) 553205
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.