Proteintech
Manufacturer Code:139631AP150UL
Catalog # 50-172-9123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KPNA5 Fusion Protein Ag4920 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 6; Importin alpha-S2; Importin subunit alpha 6 IPOA6 Karyopherin subunit alpha 5 KPNA5 SRP6; Importin subunit alpha-6; Importin subunit alpha-7; karyopherin (importin) alpha 5; karyopherin alpha 5 (importin alpha 6); Karyopherin subunit alpha-5; Karyopherin subunit alpha-6
Gene Aliases: IPOA6; IPOA7; KPNA5; Kpna6; NPI-2; SRP6
UniProt ID: (Human) O15131, (Rat) Q56R16, (Mouse) O35345
Entrez Gene ID: (Human) 3841, (Rat) 294392, (Mouse) 16650
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.