Invitrogen
Manufacturer Code:PA521033
Catalog # PIPA521033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the amino terminus of human KPNA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Importin alpha Q1; Importin subunit alpha-3; importin subunit alpha-4; IPOA3 QIP1 SRP3 importin alpha Q1 importin subunit alpha-4 karyopherin subunit alpha-4; karyopherin (importin) alpha 4; karyopherin alpha 4 (importin alpha 3); Karyopherin subunit alpha-4; predicted role in nuclear import; Qip1
Gene Aliases: 1110058D08Rik; importin; IPOA3; KPNA4; QIP1; SRP3
UniProt ID: (Human) O00629, (Mouse) O35343
Entrez Gene ID: (Human) 3840, (Mouse) 16649, (Rat) 361959
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.