Invitrogen
Manufacturer Code:PA518239
Catalog # PIPA518239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NSSANVPTEGFQF) corresponding to the C-terminus amino acids of KPNA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Importin alpha Q1; Importin subunit alpha-3; importin subunit alpha-4; karyopherin (importin) alpha 4; karyopherin alpha 4 (importin alpha 3); Karyopherin subunit alpha-4; KPNA4 IPOA3 QIP1 SRP3 MGC12217 MGC26703 karyopherin alpha 4 (importin alpha 3) importin alpha 3 importin-alpha-Q1 karyopherin alpha 4; predicted role in nuclear import; Qip1
Gene Aliases: 1110058D08Rik; importin; IPOA3; KPNA4; QIP1; SRP3
UniProt ID: (Human) O00629, (Mouse) O35343
Entrez Gene ID: (Human) 3840, (Mouse) 16649, (Rat) 361959
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.