Invitrogen
Manufacturer Code:PA518238
Catalog # PIPA518238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DPTANLQTKEFNF) corresponding to the C-terminus amino acids of KPNA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 4; Importin alpha Q2; importin alpha-3; importin subunit alpha-3; Importin subunit alpha-4; importin-alpha-Q2; karyopherin (importin) alpha 3; karyopherin alpha 3 (importin alpha 4); Karyopherin subunit alpha-3; KPNA3 IPOA4 karyopherin alpha 3 (importin alpha 4) RP11-432M24.3 SRP1gamma SRP4 hSRP1 importin alpha 4 importin alpha 4 importin-alpha-Q2 karyopherin alpha 3 SRP1 importin alpha-3; predicted role in nuclear import; Qip2; SRP1-gamma
Gene Aliases: hSRP1; importin; IPOA4; KPNA3; QIP2; SRP1; SRP1gamma; SRP4
UniProt ID: (Human) O00505, (Mouse) O35344
Entrez Gene ID: (Human) 3839, (Mouse) 16648, (Rat) 361055
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.