Abnova
Manufacturer Code:PAB19316
Catalog # 89-120-849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids near C-terminus of human KPNA3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of KPNA3.
For Research Use Only
Protein Aliases: hSRP1; importin alpha 4; Importin alpha Q2; importin alpha-3; importin subunit alpha-3; Importin subunit alpha-4; importin-alpha-Q2; IPOA4; karyopherin alpha 3 (importin alpha 4); Karyopherin subunit alpha-3; Qip2; SRP1; SRP1-gamma; SRP1gamma; SRP4
Gene Aliases: hSRP1; IPOA4; KPNA3; QIP2; SRP1; SRP1gamma; SRP4
UniProt ID: (Human) O00505
Entrez Gene ID: (Human) 3839
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.