Invitrogen
Manufacturer Code:MA517016
Catalog # PIMA517016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 2G7 |
Immunogen | Amino acids 116-515 produced in P3-x63Ag.8.653 cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Importin alpha P1; Importin subunit alpha-1; importin subunit alpha-2; importin-alpha-P1; Karyopherin A2 KPNA2 IMA2 Importin alpha 2 Importin alpha P1 Importin subunit alpha-2 IPOA1 IPOA 1 IPOA1 Karyopherin alpha 2 subunit RAG cohort 1 RCH 1 RCH1 SRP 1 SRP1 alpha SRP1; karyopherin alpha 2 (RAG cohort 1, importin alpha 1); Karyopherin subunit alpha-2; m-importin-alpha-P1; Pendulin; Pore targeting complex 58 kDa subunit; PTAC58; RAG cohort protein 1; SRP1-alpha
Gene Aliases: 2410044B12Rik; IPOA1; KPNA2; PTAC58; QIP2; RCH1; SRP1; SRP1-alpha; SRP1alpha
UniProt ID: (Human) P52292, (Mouse) P52293
Entrez Gene ID: (Human) 3838, (Mouse) 16647
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.