Invitrogen
Manufacturer Code:PA5102445
Catalog # PIPA5102445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KPNA2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410044B12Rik IMA2 importin importin alpha 2 Importin alpha P1 Importin subunit alpha-1 importin subunit alpha-2 importin-alpha-P1 IPOA 1 IPOA1 IPOA1 QIP2 karyopherin (importin) alpha 2 Karyopherin A2 karyopherin alpha 2 karyopherin alpha 2 (RAG cohort 1 importin alpha 1) karyopherin subunit alpha 2 karyopherin subunit alpha-2 Kpna2 m-importin-alpha-P1 nuclear import protein pendulin Pore targeting complex 58 kDa subunit PTAC58 QIP2 RAG cohort 1 RAG cohort protein 1 RCH 1 RCH1 SRP 1 SRP1 SRP1 alpha SRP1alpha SRP1-alpha; Importin alpha P1; Importin subunit alpha-1; importin subunit alpha-2; importin-alpha-P1; karyopherin (importin) alpha 2; karyopherin alpha 2 (RAG cohort 1, importin alpha 1); Karyopherin subunit alpha-2; m-importin-alpha-P1; nuclear import protein; Pendulin; Pore targeting complex 58 kDa subunit; PTAC58; RAG cohort protein 1; SRP1-alpha
Gene Aliases: 2410044B12Rik; importin; IPOA1; KPNA2; PTAC58; QIP2; RCH1; SRP1; SRP1-alpha; SRP1alpha
UniProt ID: (Human) P52292, (Mouse) P52293
Entrez Gene ID: (Human) 3838, (Mouse) 16647, (Rat) 85245
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.